Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027879.2 | NP_082155.1 | 9 - 10 | 1054 | 65 | |
GenBank mRNA | AK004569.1 | - | 9 - 10 | 1021 | 65 | |
AK136292.1 | - | 1 - 2 | 974 | 65 | ||
BC147503.1 | - | 9 - 10 | 1054 | 9056386 | 65 | |
BC158078.1 | - | 9 - 10 | 1054 | 9088170 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |