Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027879.2 | NP_082155.1 | 10 - 11 | 1153 | 79 | |
GenBank mRNA | AK004569.1 | - | 10 - 11 | 1120 | 79 | |
AK136292.1 | - | 2 - 3 | 1073 | 79 | ||
BC147503.1 | - | 10 - 11 | 1153 | 9056386 | 79 | |
BC158078.1 | - | 10 - 11 | 1153 | 9088170 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |