Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030250.2 | NP_084526.1 | 2 - 3 | 742 | 129 | |
GenBank mRNA | AK155620.1 | - | 2 - 3 | 743 | 129 | |
AK167741.1 | - | 2 - 3 | 731 | 129 | ||
AK167939.1 | - | 2 - 3 | 735 | 129 | ||
BC003223.1 | - | 2 - 3 | 701 | 3482163 | 129 | |
BC018372.1 | - | 2 - 3 | 717 | 3499868 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |