Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030250.2 | NP_084526.1 | 3 - 4 | 900 | 120 | |
GenBank mRNA | AK155620.1 | - | 3 - 4 | 901 | 121 | |
AK162260.1 | - | 1 - 2 | 62 | 120 | ||
AK167741.1 | - | 3 - 4 | 889 | 120 | ||
AK167939.1 | - | 3 - 4 | 893 | 120 | ||
BC003223.1 | - | 3 - 4 | 859 | 3482163 | 120 | |
BC018372.1 | - | 3 - 4 | 875 | 3499868 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |