Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008167.2 | NP_032193.1 | 7 - 8 | 1127 | 130 | |
GenBank mRNA | AK036192.1 | - | 7 - 8 | 1158 | 130 | |
AK141070.1 | - | 1979 | 130 | |||
BC139823.1 | - | 7 - 8 | 1131 | 40141838 | 130 | |
D13266.1 | - | 7 - 8 | 1127 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |