Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178675.4 | NP_848790.2 | 5 - 6 | 1290 | 84 | |
GenBank mRNA | AK038601.1 | - | 5 - 6 | 1285 | 84 | |
AK048957.1 | - | 5 - 6 | 1250 | 84 | ||
AK053426.1 | - | 5 - 6 | 1259 | 84 | ||
AK053545.1 | - | 5 - 6 | 1229 | 84 | ||
AK083768.1 | - | 5 - 6 | 1229 | 84 | ||
AK139331.1 | - | 5 - 6 | 1260 | 84 | ||
AK163806.1 | - | 5 - 6 | 1090 | 84 | ||
AK165021.1 | - | 5 - 6 | 1244 | 84 | ||
AK166268.1 | - | 5 - 6 | 1218 | 84 | ||
BC059075.1 | - | 5 - 6 | 1186 | 6826829 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |