Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172731.2 | NP_766319.2 | 11 - 12 | 3237 | 76 | |
GenBank mRNA | AK220153.1 | - | 12 - 13 | 3711 | 76 | |
BC042732.1 | - | 7 - 8 | 654 | 76 | ||
BC060664.1 | - | 11 - 12 | 2700 | 76 | ||
BC145502.1 | - | 12 - 13 | 3054 | 9054065 | 76 | |
BC150786.1 | - | 11 - 12 | 3186 | 9087697 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |