Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175314.3 | NP_780523.2 | 9 - 10 | 1169 | 59 | |
GenBank mRNA | CA325405.1 | - | 2 - 3 | 165 | 59 | |
CK637439.1 | - | 5 - 6 | 537 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mus musculus Probe spans exons |