Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001160.3 | NP_001001160.1 | 3 - 4 | 1495 | 74 | |
NM_001289674.1 | NP_001276603.1 | 2 - 3 | 1326 | 74 | ||
NM_001289675.1 | NP_001276604.1 | 2 - 3 | 1320 | 74 | ||
GenBank mRNA | AK129466.1 | - | 1 - 2 | 872 | 74 | |
AK147230.1 | - | 3 - 4 | 1387 | 74 | ||
AK147681.1 | - | 2 - 3 | 1185 | 74 | ||
BC070445.1 | - | 2 - 3 | 1326 | 30536738 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |