Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hint3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025798.3 | NP_080074.1 | 2 - 3 | 281 | 149 | |
GenBank mRNA | AK002482.1 | - | 2 - 3 | 376 | 149 | |
AK007758.1 | - | 2 - 3 | 298 | 149 | ||
AK027974.1 | - | 2 - 3 | 282 | 149 | ||
AY040768.1 | - | 2 - 3 | 284 | 149 | ||
BC025065.1 | - | 2 - 3 | 281 | 4035971 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |