Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053227.2 | NP_444457.2 | 1 | 46 | 107 | |
GenBank mRNA | BC107184.1 | - | 1 | 46 | 107 | |
BC107185.1 | - | 1 | 46 | 107 | ||
BC151088.1 | - | 1 | 143 | 9088001 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon |