Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161741.1 | NP_001155213.1 | 3 - 4 | 251 | 96 | |
GenBank mRNA | BM055959.1 | - | 2 - 3 | 199 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |