Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY036107.1 | - | 5 - 6 | 485 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |