Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Urgcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178623.3 | NP_848738.2 | 1 - 2 | 75 | 151 | |
GenBank mRNA | AK033577.1 | - | 1 - 2 | 49 | 151 | |
AK166495.1 | - | 1 - 2 | 46 | 80 | ||
BY058620.1 | - | 1 - 2 | 75 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 Mus musculus 1700 Probe spans exons |