Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpusd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033204.1 | NP_001028376.1 | 3 - 4 | 307 | 83 | |
GenBank mRNA | AK076749.1 | - | 3 - 4 | 308 | 83 | |
AK133871.1 | - | 3 - 4 | 287 | 83 | ||
BC038081.1 | - | 3 - 4 | 248 | 83 | ||
BC116825.1 | - | 3 - 4 | 307 | 40126144 | 83 | |
BC116827.1 | - | 3 - 4 | 307 | 40126146 | 83 | |
BC144733.1 | - | 3 - 4 | 307 | 9053281 | 83 | |
BC144734.1 | - | 3 - 4 | 307 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |