Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034858.3 | NP_001030030.2 | 3 - 4 | 518 | 70 | |
GenBank mRNA | BM942469.1 | - | 3 - 4 | 501 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |