Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021717.2 | NP_068363.2 | 5 - 6 | 948 | 94 | |
GenBank mRNA | AJ278170.1 | - | 5 - 6 | 971 | 94 | |
AK156526.1 | - | 5 - 6 | 821 | 94 | ||
BC115523.1 | - | 5 - 6 | 698 | 40101227 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |