Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172473.3 | NP_766061.2 | 21 - 22 | 2691 | 120 | |
GenBank mRNA | AK029162.1 | - | 11 - 12 | 1511 | 120 | |
AK042879.1 | - | 21 - 22 | 2632 | 120 | ||
AK156958.1 | - | 21 - 22 | 2633 | 120 | ||
BC025227.1 | - | 5 - 6 | 616 | 120 | ||
BC120695.1 | - | 21 - 22 | 2588 | 40129618 | 120 | |
BC120697.1 | - | 21 - 22 | 2588 | 40129620 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |