Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159389.1 | NP_001152861.1 | 6 - 7 | 718 | 114 | |
NM_177306.3 | NP_796280.1 | 3 - 4 | 349 | 114 | ||
GenBank mRNA | AK029809.1 | - | 3 - 4 | 350 | 114 | |
AK049380.1 | - | 6 - 7 | 698 | 114 | ||
FJ804414.1 | - | 6 - 7 | 668 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |