Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008548.4 | NP_032574.1 | 11 - 12 | 2240 | 87 | |
GenBank mRNA | AK031038.1 | - | 11 - 12 | 2241 | 87 | |
AK042371.1 | - | 10 - 11 | 2081 | 87 | ||
AK043932.1 | - | 11 - 12 | 2181 | 87 | ||
AK154820.1 | - | 11 - 12 | 2182 | 87 | ||
AK171841.1 | - | 11 - 12 | 2183 | 87 | ||
AK191835.1 | - | 1 - 2 | 54 | 87 | ||
AK196316.1 | - | 1 - 2 | 58 | 87 | ||
AK196883.1 | - | 1 - 2 | 134 | 87 | ||
AK197437.1 | - | 1 - 2 | 134 | 87 | ||
BC015265.1 | - | 11 - 12 | 2141 | 4223319 | 87 | |
U04299.1 | - | 10 - 11 | 1844 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |