Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175155.4 | NP_780364.3 | 13 - 14 | 1755 | 78 | |
GenBank mRNA | AJ507736.1 | - | 13 - 14 | 1570 | 78 | |
AK039638.1 | - | 1 - 2 | 94 | 78 | ||
AK129216.1 | - | 8 - 9 | 1089 | 78 | ||
AK149872.1 | - | 13 - 14 | 1707 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |