Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178440.4 | NP_848534.2 | 3 - 4 | 519 | 77 | |
GenBank mRNA | AK030825.1 | - | 3 - 4 | 520 | 77 | |
AK036461.1 | - | 3 - 4 | 504 | 77 | ||
AK042033.1 | - | 3 - 4 | 470 | 77 | ||
AK088011.1 | - | 3 - 4 | 520 | 77 | ||
AK146251.1 | - | 3 - 4 | 492 | 77 | ||
AK152797.1 | - | 3 - 4 | 513 | 77 | ||
AK159700.1 | - | 3 - 4 | 495 | 77 | ||
AK169450.1 | - | 3 - 4 | 513 | 77 | ||
AK171284.1 | - | 3 - 4 | 513 | 77 | ||
AK171505.1 | - | 3 - 4 | 492 | 77 | ||
BC113755.1 | - | 3 - 4 | 463 | 40061202 | 77 | |
BC113954.1 | - | 3 - 4 | 463 | 40061200 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |