Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008297.3 | NP_032323.3 | 3 - 4 | 455 | 85 | |
GenBank mRNA | AK052270.1 | - | 3 - 4 | 354 | 85 | |
AK053976.1 | - | 3 - 4 | 456 | 85 | ||
BC018414.1 | - | 3 - 4 | 377 | 3990765 | 85 | |
X61754.1 | - | 3 - 4 | 489 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |