Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010146.2 | NP_034276.2 | 1 - 2 | 328 | 88 | |
GenBank mRNA | AF124044.1 | - | 1 - 2 | 368 | 88 | |
BY730431.1 | - | 1 - 2 | 329 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mus musculus Probe spans exons |