Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017401.2 | NP_059097.2 | 4 - 5 | 904 | 81 | |
GenBank mRNA | AF176098.1 | - | 4 - 5 | 677 | 81 | |
AJ251804.2 | - | 4 - 5 | 648 | 81 | ||
AK137747.1 | - | 2 - 3 | 1969 | 81 | ||
BC056213.1 | - | 4 - 5 | 696 | 30024844 | 81 | |
BC056996.1 | - | 1 - 2 | 2551 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |