Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146308.1 | NP_001139780.1 | 4 - 5 | 404 | 54 | |
NM_001146309.1 | NP_001139781.1 | 4 - 5 | 404 | 54 | ||
NM_013810.3 | NP_038838.1 | 4 - 5 | 404 | 54 | ||
GenBank mRNA | AK046073.1 | - | 4 - 5 | 384 | 54 | |
AK053796.1 | - | 4 - 5 | 400 | 54 | ||
AK078082.1 | - | 4 - 5 | 371 | 54 | ||
AK134136.1 | - | 4 - 5 | 371 | 54 | ||
AK134487.1 | - | 4 - 5 | 371 | 54 | ||
AK140752.1 | - | 4 - 5 | 371 | 54 | ||
AK142818.1 | - | 4 - 5 | 354 | 54 | ||
AK146920.1 | - | 4 - 5 | 371 | 54 | ||
AK151096.1 | - | 4 - 5 | 384 | 54 | ||
AK152110.1 | - | 4 - 5 | 369 | 54 | ||
AK152204.1 | - | 4 - 5 | 384 | 54 | ||
AK153389.1 | - | 4 - 5 | 358 | 54 | ||
AK159984.1 | - | 4 - 5 | 381 | 54 | ||
AK161973.1 | - | 4 - 5 | 384 | 54 | ||
AK167956.1 | - | 4 - 5 | 344 | 54 | ||
AK168898.1 | - | 4 - 5 | 381 | 54 | ||
AK172471.1 | - | 4 - 5 | 361 | 54 | ||
AY098595.1 | - | 4 - 5 | 327 | 54 | ||
BC046430.1 | - | 4 - 5 | 369 | 4986658 | 54 | |
U58884.1 | - | 4 - 5 | 357 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |