Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159288.1 | NP_001152760.1 | 8 - 9 | 1306 | 96 | |
NM_198162.1 | NP_937805.1 | 7 - 8 | 633 | 96 | ||
GenBank mRNA | AK173042.1 | - | 8 - 9 | 1306 | 96 | |
BC059243.1 | - | 7 - 8 | 633 | 6417551 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |