Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138607.3 | NP_613073.2 | 3 - 4 | 473 | 81 | |
GenBank mRNA | AK008402.1 | - | 3 - 4 | 474 | 81 | |
BC125297.1 | - | 2 - 3 | 339 | 40129812 | 81 | |
Y18505.1 | - | 3 - 4 | 438 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |