Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008883.2 | NP_032909.2 | 23 - 24 | 4402 | 67 | |
GenBank mRNA | AJ748653.1 | - | 5 - 6 | 882 | 67 | |
AK049319.1 | - | 5 - 6 | 816 | 67 | ||
AK220285.1 | - | 19 - 20 | 3026 | 67 | ||
BC093482.1 | - | 23 - 24 | 4154 | 30945372 | 67 | |
D86950.1 | - | 23 - 24 | 4368 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |