Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290742.1 | NP_001277671.1 | 4 - 5 | 67 | ||
NM_133884.2 | NP_598645.2 | 4 - 5 | 67 | |||
GenBank mRNA | BC018407.1 | - | 4 - 5 | 4015340 | 67 | |
BQ552936.1 | - | 1 - 2 | 405 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |