Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400030418.mm.1 |
TC0400003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030418.mm.1 |
NM_001080819.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030418.mm.1 |
NM_001080819.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030417.mm.1 |
TC0400003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030417.mm.1 |
NM_001080819.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030417.mm.1 |
NM_001080819.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030420.mm.1 |
TC0400003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030420.mm.1 |
NM_001080819.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030420.mm.1 |
NM_001080819.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030421.mm.1 |
TC0400003712.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030421.mm.1 |
NM_001080819.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030421.mm.1 |
NM_001080819.1 |
Refseq |