Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285505.1 | NP_001272434.1 | 4 - 5 | 477 | 97 | |
NM_001285506.1 | NP_001272435.1 | 3 - 4 | 328 | 97 | ||
NM_009097.5 | NP_033123.2 | 4 - 5 | 477 | 97 | ||
GenBank mRNA | AK132856.1 | - | 2 - 3 | 514 | 97 | |
AK148208.1 | - | 4 - 5 | 97 | |||
AK167790.1 | - | 3 - 4 | 315 | 97 | ||
BC049076.1 | - | 4 - 5 | 428 | 97 | ||
BC094470.1 | - | 4 - 5 | 430 | 5098650 | 97 | |
BY226173.1 | - | 3 - 4 | 98 | |||
BY737295.1 | - | 4 - 5 | 462 | 97 | ||
CA547794.1 | - | 3 - 4 | 320 | 97 | ||
M28489.1 | - | 4 - 5 | 463 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |