Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tesk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146151.4 | NP_666263.3 | 9 - 10 | 1261 | 62 | |
GenBank mRNA | AK031814.1 | - | 9 - 10 | 1254 | 62 | |
AK047755.1 | - | 9 - 10 | 1262 | 62 | ||
AK134575.1 | - | 5 - 6 | 367 | 62 | ||
BC019149.1 | - | 8 - 9 | 994 | 5053345 | 62 | |
BC029766.1 | - | 9 - 10 | 1239 | 4947564 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |