Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010173.4 | NP_034303.3 | 11 - 12 | 1560 | 59 | |
GenBank mRNA | AK079598.1 | - | 11 - 12 | 1561 | 59 | |
AK080603.1 | - | 11 - 12 | 1393 | 59 | ||
AK138481.1 | - | 11 - 12 | 1180 | 59 | ||
BC006863.1 | - | 11 - 12 | 1339 | 3595191 | 59 | |
BC052321.1 | - | 11 - 12 | 1602 | 6515634 | 59 | |
U82536.1 | - | 11 - 12 | 1321 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |