Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181585.5 | NP_853616.1 | 8 - 9 | 1404 | 85 | |
GenBank mRNA | AK031936.1 | - | 8 - 9 | 1321 | 85 | |
AK132341.1 | - | 8 - 9 | 1679 | 85 | ||
AK135693.1 | - | 8 - 9 | 1713 | 85 | ||
AK161822.1 | - | 8 - 9 | 1672 | 85 | ||
AK163413.1 | - | 8 - 9 | 1678 | 85 | ||
BC053102.1 | - | 8 - 9 | 1404 | 6405534 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |