Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mknk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285487.1 | NP_001272416.1 | 11 - 12 | 994 | 86 | |
NM_001285488.1 | NP_001272417.1 | 10 - 11 | 888 | 86 | ||
NM_021461.5 | NP_067436.1 | 9 - 10 | 844 | 86 | ||
GenBank mRNA | AK155934.1 | - | 11 - 12 | 955 | 86 | |
AK169982.1 | - | 10 - 11 | 854 | 86 | ||
BC021369.1 | - | 9 - 10 | 800 | 5030044 | 86 | |
CN538557.1 | - | 1 - 2 | 112 | 86 | ||
Y11091.1 | - | 9 - 10 | 809 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |