Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd24a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009846.2 | NP_033976.1 | 1 - 2 | 152 | 58 | |
GenBank mRNA | AK133869.1 | - | 1 - 2 | 151 | 58 | |
AK168111.1 | - | 1 - 2 | 151 | 58 | ||
AK169613.1 | - | 1 - 2 | 149 | 58 | ||
BC075622.1 | - | 1 - 2 | 132 | 6415159 | 58 | |
M58661.1 | - | 1 - 2 | 139 | 58 | ||
X53825.1 | - | 1 - 2 | 90 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |