Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153055.3 | NP_694695.3 | 11 - 12 | 1317 | 94 | |
GenBank mRNA | AK049737.1 | - | 9 - 10 | 3438 | 94 | |
AY024346.1 | - | 11 - 12 | 1114 | 94 | ||
BC019366.1 | - | 11 - 12 | 1081 | 5066908 | 94 | |
BC029774.1 | - | 3 - 4 | 321 | 94 | ||
BC031846.1 | - | 11 - 12 | 1081 | 5066822 | 94 | |
BC059816.1 | - | 11 - 12 | 1316 | 94 | ||
BC080810.1 | - | 10 - 11 | 2620 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |