Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207242.2 | NP_997125.2 | 7 - 8 | 2405 | 67 | |
GenBank mRNA | AK078947.1 | - | 7 - 8 | 2306 | 67 | |
AY437866.1 | - | 7 - 8 | 2283 | 67 | ||
BC131788.1 | - | 7 - 8 | 2282 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |