Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207242.2 | NP_997125.2 | 8 - 9 | 2537 | 145 | |
GenBank mRNA | AK078947.1 | - | 8 - 9 | 2438 | 145 | |
AY437866.1 | - | 8 - 9 | 2415 | 145 | ||
BC131788.1 | - | 8 - 9 | 2414 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |