Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bves | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024285.2 | NP_077247.1 | 3 - 4 | 481 | 104 | |
GenBank mRNA | AF124510.1 | - | 2 - 3 | 342 | 104 | |
AF204174.1 | - | 3 - 4 | 479 | 104 | ||
BC132018.1 | - | 2 - 3 | 371 | 40130295 | 104 | |
BC132044.1 | - | 2 - 3 | 371 | 40130321 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |