Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290696.1 | NP_001277625.1 | 10 - 11 | 1743 | 72 | |
NM_172699.3 | NP_766287.1 | 11 - 12 | 1845 | 72 | ||
GenBank mRNA | AK082840.1 | - | 11 - 12 | 1846 | 72 | |
AK129271.1 | - | 10 - 11 | 1716 | 72 | ||
AK139508.1 | - | 11 - 12 | 1779 | 72 | ||
BC058231.1 | - | 11 - 12 | 1818 | 6314981 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |