Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110129.1 | NP_001103599.1 | 9 - 10 | 604 | 104 | |
GenBank mRNA | AK014665.1 | - | 9 - 10 | 565 | 104 | |
AK134995.1 | - | 9 - 10 | 584 | 104 | ||
AK136330.1 | - | 9 - 10 | 563 | 104 | ||
AK164299.1 | - | 9 - 10 | 569 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Inventoried Probe spans exons |