Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023906.3 | NP_076395.2 | 3 - 4 | 536 | 148 | |
GenBank mRNA | AF155354.1 | - | 2 - 3 | 351 | 148 | |
AK010315.1 | - | 3 - 4 | 472 | 148 | ||
AK028489.1 | - | 4 - 5 | 642 | 148 | ||
AK038086.1 | - | 3 - 4 | 517 | 148 | ||
AK143580.1 | - | 3 - 4 | 471 | 148 | ||
BC023086.1 | - | 3 - 4 | 470 | 4947610 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |