Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025745.3 | NP_080021.3 | 1 - 2 | 222 | 131 | |
GenBank mRNA | AK002471.1 | - | 1 - 2 | 213 | 131 | |
AK077120.1 | - | 1 - 2 | 215 | 131 | ||
AK133874.1 | - | 1 - 2 | 223 | 131 | ||
BC018468.1 | - | 1 - 2 | 232 | 3591901 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |