Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400013578.mm.1 |
NM_001285482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013578.mm.1 |
NM_008309.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013578.mm.1 |
TC0400001686.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013581.mm.1 |
NM_001285482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013581.mm.1 |
NM_001285483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013581.mm.1 |
NM_001285484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013581.mm.1 |
NM_008309.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013581.mm.1 |
TC0400001686.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400013582.mm.1 |
NM_001285482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013582.mm.1 |
NM_001285483.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013582.mm.1 |
NM_001285484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013582.mm.1 |
NM_008309.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400013582.mm.1 |
TC0400001686.mm.1 |
MTA TC |