Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019684.1 | NP_062658.1 | 11 - 12 | 1275 | 99 | |
GenBank mRNA | AF043288.1 | - | 11 - 12 | 1275 | 99 | |
AJ748651.1 | - | 10 - 11 | 1099 | 99 | ||
AK004696.2 | - | 8 - 9 | 1944 | 99 | ||
BC120883.1 | - | 11 - 12 | 1270 | 40057934 | 99 | |
BC120884.1 | - | 11 - 12 | 1270 | 40057937 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |