Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177973.1 | NP_001171444.1 | 9 - 10 | 1410 | 67 | |
NM_001177974.1 | NP_001171445.1 | 9 - 10 | 1407 | 67 | ||
NM_001177975.1 | NP_001171446.1 | 9 - 10 | 1407 | 67 | ||
NM_001177976.1 | NP_001171447.1 | 9 - 10 | 1407 | 67 | ||
NM_008363.2 | NP_032389.2 | 9 - 10 | 1407 | 67 | ||
GenBank mRNA | AB088370.1 | - | 9 - 10 | 1234 | 67 | |
AF103876.1 | - | 9 - 10 | 1407 | 67 | ||
AK045958.1 | - | 9 - 10 | 1316 | 67 | ||
AK052933.1 | - | 7 - 8 | 1277 | 67 | ||
AK143694.1 | - | 9 - 10 | 1318 | 67 | ||
AK144879.1 | - | 9 - 10 | 1316 | 67 | ||
AK196997.1 | - | 1 - 2 | 94 | 67 | ||
AY184362.1 | - | 9 - 10 | 1407 | 67 | ||
AY184363.1 | - | 9 - 10 | 1407 | 67 | ||
AY184364.1 | - | 9 - 10 | 1407 | 67 | ||
BC004778.1 | - | 9 - 10 | 1402 | 3257579 | 67 | |
U56773.2 | - | 9 - 10 | 1231 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |