Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019587.2 | NP_062533.2 | 6 - 7 | 1645 | 65 | |
GenBank mRNA | AK122467.1 | - | 5 - 6 | 571 | 65 | |
AK138168.1 | - | 5 - 6 | 598 | 65 | ||
AK138631.1 | - | 6 - 7 | 720 | 65 | ||
AK140240.1 | - | 6 - 7 | 1649 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |