Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142809.1 | NP_001136281.1 | 10 - 11 | 2154 | 76 | |
NM_001142810.1 | NP_001136282.1 | 10 - 11 | 2145 | 76 | ||
NM_133987.2 | NP_598748.1 | 10 - 11 | 2169 | 76 | ||
GenBank mRNA | AB077327.1 | - | 10 - 11 | 1502 | 76 | |
AF459435.1 | - | 10 - 11 | 2152 | 76 | ||
AJ748650.1 | - | 7 - 8 | 744 | 76 | ||
BC024444.1 | - | 9 - 10 | 1167 | 76 | ||
BC029000.1 | - | 8 - 9 | 917 | 76 | ||
BC049801.1 | - | 10 - 11 | 1320 | 76 | ||
BC141067.1 | - | 11 - 12 | 1895 | 9055679 | 76 | |
BC145319.1 | - | 11 - 12 | 1559 | 9053874 | 76 | |
EU523148.1 | - | 6 - 7 | 1203 | 76 | ||
KC994641.1 | - | 5 - 6 | 1065 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |